1-(4-Methoxyphenyl)cyclopropancarbonsäure 97%

Lieferant: Apollo Scientific

OR61139-5G
APOSOR61139-5GEA 65.4 EUR
APOSOR61139-5G
1-(4-Methoxyphenyl)cyclopropancarbonsäure 97%
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid
Formel: C₁₁H₁₂O₃
Schmelzpunkt: 130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00019223
CAS-Nummer: 16728-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR