1-(3-Methoxyphenyl)cyclobutanecarboxylic acid

Lieferant: Apollo Scientific
Achtung

OR61141-1G
APOSOR61141-1GEA 348 EUR
APOSOR61141-1G
1-(3-Methoxyphenyl)cyclobutanecarboxylic acid
1-(3-Methoxyphenyl)cyclobutanecarboxylic acid
Formel: C₁₂H₁₄O₃
Molecular Weight: 206,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09414711
CAS-Nummer: 74205-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR