4-Chloro-3-hydroxybenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR6115-250MG
APOSOR6115-250MGEA
21.9
EUR
APOSOR6115-250MG
4-Chloro-3-hydroxybenzoic acid
4-Chloro-3-hydroxybenzoic acid
Formel:
C₇H₅ClO₃ Molecular Weight: 172,5700 g/mol Schmelzpunkt: 216…218 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153892 CAS-Nummer: 34113-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...