2-Bromo-4,5,6,7-tetrahydrobenzo[b]thiophene

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-4,5,6,7-tetrahydro-1-benzothiophene

OR61152-1G
APOSOR61152-1GEA 290 EUR
APOSOR61152-1G
2-Bromo-4,5,6,7-tetrahydrobenzo[b]thiophene
2-Bromo-4,5,6,7-tetrahydrobenzo[b]thiophene
Formel: C₈H₉BrS
Molecular Weight: 217,126 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20037421
CAS-Nummer: 111873-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR