2-(3-Bromothien-2-yl)-1,3,4-oxadiazole

Lieferant: Apollo Scientific

OR61154-1G
APOSOR61154-1GEA 319 EUR
APOSOR61154-1G
2-(3-Bromothien-2-yl)-1,3,4-oxadiazole
2-(3-Bromothien-2-yl)-1,3,4-oxadiazole
Formel: C₆H₃BrN₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD20037422
CAS-Nummer: 1357064-43-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR