4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR61155-1G
APOSOR61155-1GEA
342
EUR
APOSOR61155-1G
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulphonyl chloride
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-sulphonyl chloride
Formel:
C₈H₉ClO₂S₂ Schmelzpunkt: 64…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18825753 CAS-Nummer: 128852-17-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...