2-(4-Bromophenyl)-2-azaspiro[4.4]nonane-1,3-dione

Lieferant: Apollo Scientific

Synonyme: 2-(4-Bromophenyl)-1,3-dioxo-2-azaspiro[4.4]nonane, 1-Bromo-4-(1,3-dioxo-2-azaspiro[4.4]non-2-yl)benzene

OR61157-1G
APOSOR61157-1GEA 290 EUR
APOSOR61157-1G
2-(4-Bromophenyl)-2-azaspiro[4.4]nonane-1,3-dione
2-(4-Bromophenyl)-2-azaspiro[4.4]nonane-1,3-dione
Formel: C₁₄H₁₄BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01763484
CAS-Nummer: 61343-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR