6-(1H-Imidazol-1-yl)nicotinic acid

Lieferant: Apollo Scientific
Achtung

OR6116-1G OR6116-5G
APOSOR6116-1GEA 170 EUR
APOSOR6116-1G APOSOR6116-5G
6-(1H-Imidazol-1-yl)nicotinic acid
6-(1H-Imidazol-1-yl)nicotinic acid
Formel: C₉H₇N₃O₂
Molecular Weight: 189.18 g/mol
Schmelzpunkt: >270 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00140821
CAS-Nummer: 216955-75-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR