3-Bromo-2-nitrobenzonitrile 95%

Lieferant: Apollo Scientific
Achtung

OR61169-1G OR61169-5G OR61169-500MG
APOSOR61169-1GEA 102 EUR
APOSOR61169-1G APOSOR61169-5G APOSOR61169-500MG
3-Bromo-2-nitrobenzonitrile 95%
3-Bromo-2-nitrobenzonitrile
Formel: C₇H₃BrN₂O₂
Molecular Weight: 227.015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18392276
CAS-Nummer: 1261777-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR