2-Amino-3-pyridinemethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Amino-pyridin-3-yl)-methanol, 3-(Hydroxymethyl)-2-pyridinamine, 2-Amino-3-pyridinemethanol, 2-Amino-3-hydroxymethylpyridine, 2-Amino-3-pyridylmethanol, 2-Amino-3-(hydroxymethyl)pyridine, 2-Aminopyridine-3-methanol
OR6117-1G
APOSOR6117-1GEA
24.8
EUR
APOSOR6117-1G
2-Amino-3-pyridinemethanol
2-Amino-3-pyridinemethanol
Formel:
C₆H₈N₂O Molecular Weight: 124,14 g/mol Siedepunkt: 279 °C Schmelzpunkt: 138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663510 CAS-Nummer: 23612-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...