3-Aminopyrrolidine, 3-BOC protected hydrochloride 95+%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (pyrrolidin-3-yl)carbamate hydrochloride, 3-[(tert-Butoxycarbonyl)amino]pyrrolidine hydrochloride, tert-Butyl pyrrolidin-3-ylcarbamate hydrochloride

OR61180-5G
APOSOR61180-5GEA 95.7 EUR
APOSOR61180-5G
3-Aminopyrrolidine, 3-BOC protected hydrochloride 95+%
3-Aminopyrrolidine, 3-BOC protected hydrochloride
Formel: C₉H₁₈N₂O₂·ClH
Molecular Weight: 222,712 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11101358
CAS-Nummer: 1188263-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR