3-Bromo-5-chloro-2-iodobenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Bromo-5-chloro-2-iodophenyl)methanol

OR61184-1G
APOSOR61184-1GEA 609 EUR
APOSOR61184-1G
3-Bromo-5-chloro-2-iodobenzyl alcohol 95%
3-Bromo-5-chloro-2-iodobenzyl alcohol
Formel: C₇H₅BrClIO
Molecular Weight: 347.38 g/mol
MDL: MFCD30573894
CAS-Nummer: 213771-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR