2-Bromo-3-chloro-4-iodopyridine

Lieferant: Apollo Scientific
Achtung

OR61192-1G
APOSOR61192-1GEA 406 EUR
APOSOR61192-1G
2-Bromo-3-chloro-4-iodopyridine
2-Bromo-3-chloro-4-iodopyridine
Formel: C₅H₂BrClIN
Molecular Weight: 318,338 g/mol
MDL: MFCD28734063
CAS-Nummer: 1805015-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR