4-Chlorphenylharnstoff 98%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Chlorophenyl)urea

OR61200-25G
APOSOR61200-25GEA 154 EUR
APOSOR61200-25G
4-Chlorphenylharnstoff 98%
4-Chlorphenylharnstoff
Formel: C₇H₇ClN₂O
Molecular Weight: 170,6 g/mol
Schmelzpunkt: 205…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014788
CAS-Nummer: 140-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR