4-(Methylsulphonyl)-2-nitrophenol 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Hydroxy-5-(methylsulphonyl)nitrobenzene, 4-Hydroxy-3-nitrophenyl methyl sulphone

OR61203-1G OR61203-5G OR61203-25G
APOSOR61203-1GEA 23.2 EUR
APOSOR61203-1G APOSOR61203-5G APOSOR61203-25G
4-(Methylsulphonyl)-2-nitrophenol 95+%
4-(Methylsulphonyl)-2-nitrophenol
Formel: C₇H₇NO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD00085946
CAS-Nummer: 97-10-9
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR