2-(Benzhydrylthio)acetamide 98%

Lieferant: Apollo Scientific

Synonyme: 2-[(Diphenylmethyl)thio]acetamide, 2-[(Diphenylmethyl)sulphanyl]acetamide, 2-(Benzhydrylsulphanyl)acetamide

OR61208-25G
APOSOR61208-25GEA 98.6 EUR
APOSOR61208-25G
2-(Benzhydrylthio)acetamide 98%
2-(Benzhydrylthio)acetamide
Formel: C₁₅H₁₅NOS
Molecular Weight: 257,351 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00876107
CAS-Nummer: 68524-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR