2-(Cyclohexyloxy)isonicotinonitrile

Lieferant: Apollo Scientific

OR61211-1G
APOSOR61211-1GEA 149 EUR
APOSOR61211-1G
2-(Cyclohexyloxy)isonicotinonitrile
2-(Cyclohexyloxy)isonicotinonitrile
Formel: C₁₂H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09942305
CAS-Nummer: 1016867-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR