2-Amino-4-(4-nitrophenyl)thiazol 97%

Lieferant: Apollo Scientific

OR6122-1G
APOSOR6122-1GEA 139 EUR
APOSOR6122-1G
2-Amino-4-(4-nitrophenyl)thiazol 97%
2-Amino-4-(4-nitrophenyl)thiazole
Formel: C₉H₇N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00170222
CAS-Nummer: 2104-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR