1-(2-Bromophenyl)cyclopropane-1-carboxylic acid 97%
Lieferant: Apollo Scientific
Synonyme:
1-Carboxy-1-(2-bromophenyl)cyclopropane, 1-(2-Bromophenyl)cyclopropanecarboxylic acid, 1-(1-Carboxycycloprop-1-yl)-2-bromobenzene
OR61220-1G
APOSOR61220-1GEA
82.8
EUR
APOSOR61220-1G
1-(2-Bromophenyl)cyclopropane-1-carboxylic acid 97%
1-(2-Bromophenyl)cyclopropane-1-carboxylic acid
Formel:
C₁₀H₉BrO₂ Molecular Weight: 241,081 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07374434 CAS-Nummer: 124276-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...