2-Acetyl-4-chloro-2H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Chloro-2H-indazol-2-yl)ethan-1-one
OR61230-1G
APOSOR61230-1GEA
192
EUR
APOSOR61230-1G
2-Acetyl-4-chloro-2H-indazole
2-Acetyl-4-chloro-2H-indazole
Formel:
C₉H₇ClN₂O Molecular Weight: 194,618 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14583142 CAS-Nummer: 1303890-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...