2-Butyl-5-nitrobenzoxazol

Lieferant: Apollo Scientific

OR61231-1G OR61231-10G OR61231-5G
APOSOR61231-1GEA 0 EUR
APOSOR61231-1G APOSOR61231-10G APOSOR61231-5G
2-Butyl-5-nitrobenzoxazol
2-Butyl-5-nitrobenzoxazole
Formel: C₁₁H₁₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05663810
CAS-Nummer: 886360-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR