1-(3-Bromophenyl)cyclopropan-1-amine, N-BOC protected 97%

Lieferant: Apollo Scientific

Synonyme: 1-(3-Bromophenyl)-1-[(tert-butoxycarbonyl)amino]cyclopropane, tert-Butyl [1-(3-bromophenyl)cycloprop-1-yl]carbamate, tert-Butyl (1-(3-bromophenyl)cyclopropyl)carbamate

OR61242-250MG
APOSOR61242-250MGEA 241 EUR
APOSOR61242-250MG
1-(3-Bromophenyl)cyclopropan-1-amine, N-BOC protected 97%
1-(3-Bromophenyl)cyclopropan-1-amine, N-BOC protected
Formel: C₁₄H₁₈BrNO₂
Molecular Weight: 312,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19381711
CAS-Nummer: 597563-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR