1-(3-Bromophenyl)cyclopropan-1-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Bromophenyl)cyclopropanamine hydrochloride

OR61243-1G
APOSOR61243-1GEA 519 EUR
APOSOR61243-1G
1-(3-Bromophenyl)cyclopropan-1-amine hydrochloride
1-(3-Bromophenyl)cyclopropan-1-amine hydrochloride
Formel: C₉H₁₀BrN·ClH
Molecular Weight: 248,547 g/mol
Schmelzpunkt: 202…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11616353
CAS-Nummer: 597563-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR