3-(4-Bromophenyl)oxetan-3-amine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-3-(4-bromophenyl)oxetane hydrochloride, 2-(4-Bromophenyl)-1,3-epoxypropan-2-amine hydrochloride
OR61249-1G
APOSOR61249-1GEA
1290
EUR
APOSOR61249-1G
3-(4-Bromophenyl)oxetan-3-amine hydrochloride
3-(4-Bromophenyl)oxetan-3-amine hydrochloride
Formel:
C₉H₁₀BrNO·ClH Molecular Weight: 264,547 g/mol Schmelzpunkt: 234…236 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD19687804 CAS-Nummer: 1349718-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...