4-Chloro-α,α-dimethylphenethyl alcohol 96%
Lieferant: Apollo Scientific
Synonyme:
1-(Chlorophenyl)-2-hydroxy-2-methylpropane, 1-(4-Chlorophenyl)-2-methylpropan-2-ol
OR61252-1G
APOSOR61252-1GEA
186
EUR
APOSOR61252-1G
4-Chloro-α,α-dimethylphenethyl alcohol 96%
4-Chloro-α,α-dimethylphenethyl alcohol
Formel:
C₁₀H₁₃ClO Molecular Weight: 184,663 g/mol Schmelzpunkt: 34 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004466 CAS-Nummer: 5468-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...