7-Chloro-6-nitro-4-hydroxyquinazoline 95+%
Lieferant: Apollo Scientific
Synonyme:
7-Chloro-6-nitroquinazolin-4(3H)-one, 7-Chloro-6-nitro-4(3H)-quinazolinone, 7-Chloro-6-nitro-4-quinazolinol, 7-Chloro-6-nitro-4-quinazolinone
OR61256-25G
APOSOR61256-25GEA
63.8
EUR
APOSOR61256-25G
7-Chloro-6-nitro-4-hydroxyquinazoline 95+%
7-Chloro-6-nitro-4-hydroxyquinazoline
Formel:
C₈H₄ClN₃O₃ Molecular Weight: 225,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09388771 CAS-Nummer: 53449-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...