1,5,6,7-Tetrahydro-4H-indol-4-on

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Oxo-4,5,6,7-tetrahydroindole, 6,7-Dihydro-1H-indol-4(5H)-one

OR61260-25G
APOSOR61260-25GEA 77 EUR
APOSOR61260-25G
1,5,6,7-Tetrahydro-4H-indol-4-on
1,5,6,7-Tetrahydro-4H-indol-4-one
Formel: C₈H₉NO
Molecular Weight: 135.17 g/mol
Schmelzpunkt: 190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00075438
CAS-Nummer: 13754-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR