(S)-N-tert-Butoxycarbonylalanine methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Boc-Ala-OMe, N-Boc-L-alanine methyl ester

OR61274-100G
APOSOR61274-100GEA 122 EUR
APOSOR61274-100G
(S)-N-tert-Butoxycarbonylalanine methyl ester 95%
(S)-N-tert-Butoxycarbonylalanine methyl ester
Formel: C₉H₁₇NO₄
Molecular Weight: 203,24 g/mol
Schmelzpunkt: 32…35 °C
Dichte: 1,03 g/cm³ (25 °C)
Flammpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038513
CAS-Nummer: 28875-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR