3,4-Dichlorostyrene 95+%

Lieferant: Apollo Scientific

Synonyme: 1,2-Dichloro-4-ethenylbenzene, 1,2-Dichloro-4-vinylbenzene

OR61295-250MG
APOSOR61295-250MGEA 26.1 EUR
APOSOR61295-250MG
3,4-Dichlorostyrene 95+%
3,4-Dichlorostyrene
Formel: C₈H₆Cl₂
Molecular Weight: 173.039 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013625
CAS-Nummer: 2039-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR