6-Chloroimidazo[1,2-a]pyrimidine 98%

Lieferant: Apollo Scientific

OR61303-250MG
APOSOR61303-250MGEA 21.9 EUR
APOSOR61303-250MG
6-Chloroimidazo[1,2-a]pyrimidine 98%
6-Chloroimidazo[1,2-a]pyrimidine
Formel: C₆H₄ClN₃
Lagertemperatur: Kühlschrank
MDL: MFCD10574899
CAS-Nummer: 944906-56-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR