tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl bis(2-hydroxyethyl)carbamate, N-BOC-Diethanolamine

OR61309-100G
APOSOR61309-100GEA 98.6 EUR
APOSOR61309-100G
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate 97%
tert-Butyl-N,N-bis(2-hydroxyethyl)carbamate
Formel: C₉H₁₉NO₄
Molecular Weight: 205.26 g/mol
Dichte: 1.085 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD01862953
CAS-Nummer: 103898-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR