4-Formyl-3-hydroxybenzonitrile 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Cyano-2-formylphenol, 4-Cyano-2-hydroxybenzaldehyde
OR61317-250MG
OR61317-1G
APOSOR61317-250MGEA
42.2
EUR
APOSOR61317-250MG
APOSOR61317-1G
4-Formyl-3-hydroxybenzonitrile 95+%
4-Formyl-3-hydroxybenzonitrile
Formel:
C₈H₅NO₂ Schmelzpunkt: 124…125 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08361770 CAS-Nummer: 84102-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...