N-Boc-O-methyl-L-threonine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S,3R)-2-(Boc-amino)-3-methoxybutyric acid, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-methoxybutanoic acid, Boc-Thr(Me)-OH

OR61332-1G OR61332-25G OR61332-5G
APOSOR61332-1GEA 21.9 EUR
APOSOR61332-1G APOSOR61332-25G APOSOR61332-5G
N-Boc-O-methyl-L-threonine 98%
N-Boc-O-methyl-L-threonine
Formel: C₁₀H₁₉NO₅
Molecular Weight: 233.3 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00076990
CAS-Nummer: 48068-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR