1,4-Bis(2-methylstyryl)benzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Bis-MSB, 1,4-Di(2-methylstyryl)benzene, 1-Methyl-2-(2-(4-[2-(2-methylphenyl)ethenyl]phenyl)ethenyl)benzene, p-Bis(o-Methylstyryl) Benzene
OR61337-25G
OR61337-5G
OR61337-100G
APOSOR61337-25GEA
87
EUR
APOSOR61337-25G
APOSOR61337-5G
APOSOR61337-100G
1,4-Bis(2-methylstyryl)benzene
1,4-Bis(2-methylstyryl)benzene
Formel:
C₂₄H₂₂ Molecular Weight: 310.44 g/mol Schmelzpunkt: 180…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008529 CAS-Nummer: 13280-61-0 EINECS: 236-285-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...