1,4-Bis(2-methylstyryl)benzene

Lieferant: Apollo Scientific
Achtung

Synonyme: Bis-MSB, 1,4-Di(2-methylstyryl)benzene, 1-Methyl-2-(2-(4-[2-(2-methylphenyl)ethenyl]phenyl)ethenyl)benzene, p-Bis(o-Methylstyryl) Benzene

OR61337-25G OR61337-5G OR61337-100G
APOSOR61337-25GEA 87 EUR
APOSOR61337-25G APOSOR61337-5G APOSOR61337-100G
1,4-Bis(2-methylstyryl)benzene
1,4-Bis(2-methylstyryl)benzene
Formel: C₂₄H₂₂
Molecular Weight: 310.44 g/mol
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008529
CAS-Nummer: 13280-61-0
EINECS: 236-285-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR