3-Amino-1-N-Cbz-piperidine 97%

Lieferant: Apollo Scientific

Synonyme: 3-Aminopiperidine, N1-CBZ protected

OR6134-10G OR6134-25G OR6134-5G
APOSOR6134-10GEA 139 EUR
APOSOR6134-10G APOSOR6134-25G APOSOR6134-5G
3-Amino-1-N-Cbz-piperidine 97%
3-Amino-1-N-Cbz-piperidine
Formel: C13H18N2O2
Lagertemperatur: Raumtemperatur
CAS-Nummer: 711002-74-3
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR