(1R,5S,6R)-(3-Azabicyclo[3.1.0]hex-6-yl)methanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
exo-3-Azabicyclo[3.1.0]hexan-6-ylmethanol, (1R,5S,6R)-6-(Hydroxymethyl)-3-azabicyclo[3.1.0]hexane
OR61350-1G
APOSOR61350-1GEA
611
EUR
APOSOR61350-1G
(1R,5S,6R)-(3-Azabicyclo[3.1.0]hex-6-yl)methanol 95%
(1R,5S,6R)-(3-Azabicyclo[3.1.0]hex-6-yl)methanol
Formel:
C₆H₁₁NO Schmelzpunkt: 118…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11099879 CAS-Nummer: 134575-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...