Methyl-5-amino-2-(3-{[(tert-butoxy)carbonyl]amino}piperidin-1-yl)benzoate 95+%
Lieferant: Apollo Scientific
OR61360-1G
OR61360-500MG
APOSOR61360-1GEA
671
EUR
APOSOR61360-1G
APOSOR61360-500MG
Methyl-5-amino-2-(3-{[(tert-butoxy)carbonyl]amino}piperidin-1-yl)benzoate 95+%
Methyl-5-amino-2-(3-{[(tert-butoxy)carbonyl]amino}piperidin-1-yl)benzoate
Formel:
C₁₈H₂₇N₃O₄ Schmelzpunkt: <50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18803532 CAS-Nummer: 1280787-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...