2,5-Dichloronicotinonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR61381-5G
APOSOR61381-5GEA
194
EUR
APOSOR61381-5G
2,5-Dichloronicotinonitrile 97%
2,5-Dichloronicotinonitrile
Formel:
C₆H₂Cl₂N₂ Schmelzpunkt: 121…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185481 CAS-Nummer: 126954-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...