2-(4-Formyl-3-methoxyphenoxy)acetic acid

Lieferant: Apollo Scientific
Achtung

OR61394-1G OR61394-5G
APOSOR61394-1GEA 0 EUR
APOSOR61394-1G APOSOR61394-5G
2-(4-Formyl-3-methoxyphenoxy)acetic acid
2-(4-Formyl-3-methoxyphenoxy)acetic acid
Formel: C₁₀H₁₀O₅
Schmelzpunkt: 207…209 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00058468
CAS-Nummer: 84969-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR