4-[5-(Methoxycarbonyl)pyridin-2-yl]piperazine, N1-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-[5-(methoxycarbonyl)pyridin-2-yl]piperazine-1-carboxylate, Methyl 6-[4-(tert-butoxycarbonyl)piperazin-1-yl]nicotinate
OR61397-1G
APOSOR61397-1GEA
174
EUR
APOSOR61397-1G
4-[5-(Methoxycarbonyl)pyridin-2-yl]piperazine, N1-BOC protected
4-[5-(Methoxycarbonyl)pyridin-2-yl]piperazine, N1-BOC protected
Formel:
C₁₆H₂₃N₃O₄ Molecular Weight: 321,372 g/mol Schmelzpunkt: 136…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD19381738 CAS-Nummer: 857284-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...