4-Chloro-7-(morpholin-4-yl)quinazoline

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(4-Chloroquinazolin-7-yl)morpholine, 4-Chloro-7-(morpholin-4-yl)-1,3-benzodiazine

OR61399-1G
APOSOR61399-1GEA 151 EUR
APOSOR61399-1G
4-Chloro-7-(morpholin-4-yl)quinazoline
4-Chloro-7-(morpholin-4-yl)quinazoline
Formel: C₁₂H₁₂ClN₃O
Molecular Weight: 249,696 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24850028
CAS-Nummer: 1334602-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR