1,2,3,5-Tetra-O-benzoyl-2-C-methyl-β-D-ribofuranose 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2S,3R,4R,5R)-2,4-bis(benzoyloxy)-5-[(benzoyloxy)methyl]-3-methyloxolan-3-yl benzoate, (2S,3R,4R,5R)-5-((Benzoyloxy)methyl)-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate
OR61418-5G
APOSOR61418-5GEA
75.4
EUR
APOSOR61418-5G
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-β-D-ribofuranose 98%
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose
Formel:
C₃₄H₂₈O₉ Molecular Weight: 580,589 g/mol Schmelzpunkt: 154…155 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD07369658 CAS-Nummer: 15397-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...