(S)-N-Boc-2-hydroxymethylmorpholine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-tert-Butyl 2-(hydroxymethyl)morpholine-4-carboxylate, (2S)-2-(Hydroxymethyl)morpholine, N-BOC protected

OR61432-1G
APOSOR61432-1GEA 21.9 EUR
APOSOR61432-1G
(S)-N-Boc-2-hydroxymethylmorpholine 97%
(S)-N-Boc-2-hydroxymethylmorpholine
Formel: C₁₀H₁₉NO₄
Molecular Weight: 217.265 g/mol
Schmelzpunkt: 60…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09260606
CAS-Nummer: 135065-76-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR