4,6-Dichloropyridin-3-amine 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2,4-dichloropyridine

OR61439-5G
APOSOR61439-5GEA 162 EUR
APOSOR61439-5G
4,6-Dichloropyridin-3-amine 97+%
4,6-Dichloropyridin-3-amine
Formel: C₅H₄Cl₂N₂
Molecular Weight: 163.001 g/mol
Schmelzpunkt: 80…85 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00234054
CAS-Nummer: 7321-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR