2-Methyl-2-propanyl-(2-hydroxyethyl)methylcarbamat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(tert-Butoxycarbonyl)-N-methylethanolamine, N-Boc-N-methyl-2-aminoethanol, tert-Butyl (2-Hydroxyethyl)(methyl)carbamate, N-(tert-Butoxycarbonyl)-N-methyl-2-aminoethanol, N-Boc-N-methylethanolamine, N-Boc-N-methylglycinol, N-(tert-Butoxycarbonyl)-N-methylglycinol, (2-Hydroxyethyl)(methyl)carbamic acid tert-Butyl ester

OR61447-10G OR61447-100G OR61447-25G OR61447-5G
APOSOR61447-10GEA 94.4 EUR
APOSOR61447-10G APOSOR61447-100G APOSOR61447-25G APOSOR61447-5G
2-Methyl-2-propanyl-(2-hydroxyethyl)methylcarbamat 97%
tert-Butyl (2-hydroxyethyl)(methyl)carbamate
Formel: C₈H₁₇NO₃
Siedepunkt: 83…84 °C/0,05mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD03425857
CAS-Nummer: 57561-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR