cis-4-Amino-1-methylcyclohexanol 97%

Lieferant: Apollo Scientific
Gefahr

OR61449-1G OR61449-5G
APOSOR61449-1GEA 163 EUR
APOSOR61449-1G APOSOR61449-5G
cis-4-Amino-1-methylcyclohexanol 97%
cis-4-Amino-1-methylcyclohexanol
Formel: C₇H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18791201
CAS-Nummer: 177906-46-6
UN: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR