2-Bromo-5-iodobenzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Bromo-5-iodophenyl)methanol
OR61452-5G
OR61452-25G
APOSOR61452-5GEA
21.9
EUR
APOSOR61452-5G
APOSOR61452-25G
2-Bromo-5-iodobenzyl alcohol
2-Bromo-5-iodobenzyl alcohol
Formel:
C₇H₆BrIO Molecular Weight: 312.93 g/mol Schmelzpunkt: 112…116 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD11044842 CAS-Nummer: 946525-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...