(S)-(-)-3-Aminochinuclidin Dihydrochlorid 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-Quinuclidin-3-amine dihydrochloride, (S)-Quinuclidin-3-amine 2HCl, (S)-(-)-3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

OR6149-25G
APOSOR6149-25GEA 191 EUR
APOSOR6149-25G
(S)-(-)-3-Aminochinuclidin Dihydrochlorid 99%
(S)-(-)-3-Aminochinuclidin Dihydrochlorid
Formel: C₇H₁₆Cl₂N₂
Molecular Weight: 199.1213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191753
CAS-Nummer: 119904-90-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR