1-Benzhydrylazetidin-3-amine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-1-(diphenylmethyl)azetidine

OR6150-1G
APOSOR6150-1GEA 21.9 EUR
APOSOR6150-1G
1-Benzhydrylazetidin-3-amine 97%
1-Benzhydrylazetidin-3-amine
Formel: C₁₆H₁₈N₂
Molecular Weight: 238.34 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03093386
CAS-Nummer: 40432-52-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR